Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KDELR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006801.2 | NP_006792.1 | 4 | 788 | ||
GenBank mRNA | AB097047.1 | - | 4 | 786 | ||
AK075185.1 | - | 3 | 833 | |||
AK312647.1 | - | 4 | 800 | |||
BC018778.1 | - | 4 | 788 | 4870476 | ||
X55885.1 | - | 4 | 739 |