Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042595.2 | NP_001036060.1 | 4 | 871 | ||
NM_001098821.1 | NP_001092291.1 | 4 | 979 | |||
NM_001098822.1 | NP_001092292.1 | 4 | 831 | |||
NM_001098823.1 | NP_001092293.1 | 4 | 820 | |||
NM_001098824.1 | NP_001092294.1 | 4 | 808 | |||
NM_001098825.1 | NP_001092295.1 | 4 | 639 | |||
GenBank mRNA | AK130820.1 | - | 4 | 1288 | ||
AL833095.1 | - | 3 | 1742 | |||
BC063705.1 | - | 4 | 843 | 4804092 | ||
BX090160.1 | - | 3 | 514 |