Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRELID3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256403.1 | NP_001243332.1 | 4 | 435 | ||
NM_016045.2 | NP_057129.2 | 5 | 525 | |||
GenBank mRNA | AF151865.1 | - | 5 | 493 | ||
AK002157.1 | - | 5 | 468 | |||
AK298135.1 | - | 4 | 435 | |||
AK314449.1 | - | 5 | 465 | |||
BC010649.1 | - | 5 | 477 | 3873112 | ||
BC013969.2 | - | 5 | 491 | 3631601 |
Gene Symbol: SLMO2-ATP5E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037929.1 | 5 | 525 | |||
NR_037930.1 | 3 | 266 | ||||
GenBank mRNA | CD522026.1 | - | 3 | 205 | ||
FY212904.1 | - | 5 | 414 |