Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC-PINT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034120.1 | 3 | 472 | |||
NR_110472.1 | 3 | 472 | ||||
NR_110473.1 | 3 | 472 | ||||
GenBank mRNA | AK095549.1 | - | 3 | 475 | ||
DA182364.1 | - | 5 | 446 | |||
GU321462.1 | - | 3 | 293 | |||
GU321464.1 | - | 2 | 93 | |||
GU321466.1 | - | 2 | 93 |