Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000812.3 | NP_000803.2 | 7 | 1073 | ||
XM_017007985.1 | XP_016863474.1 | 3 | 211 | |||
GenBank mRNA | AK225167.1 | - | 7 | 750 | ||
AK225686.1 | - | 7 | 777 | |||
AK293691.1 | - | 6 | 650 | |||
AK312720.1 | - | 7 | 719 | |||
BC022449.1 | - | 7 | 777 | 4797401 | ||
KJ534841.1 | - | 7 | 699 | |||
KJ535054.1 | - | 6 | 631 | |||
X14767.1 | - | 7 | 730 |