Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CEP170 SIRNA Assays ›
Gene Symbol:
CEP170
Gene Name:
centrosomal protein 170
Gene Aliases:
FAM68A, KAB, KIAA0470
Chromosome Location:
Chr.1: 243124428 - 243255936 on Build GRCh38
UniGene ID:
Hs.533635
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s54935
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CEP170
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001042404.1
NP_001035863.1
12
2205
NM_001042405.1
NP_001035864.1
12
2205
NM_014812.2
NP_055627.2
13
2499
XM_006711843.3
XP_006711906.1
12
3208
XM_011544334.2
XP_011542636.1
12
3215
XM_011544335.2
XP_011542637.1
12
2628
XM_011544336.2
XP_011542638.1
12
3534
XM_011544337.2
XP_011542639.1
12
3215
XM_011544338.2
XP_011542640.1
12
3215
XM_011544339.2
XP_011542641.1
12
3215
XM_011544340.2
XP_011542642.1
12
3165
XM_011544341.2
XP_011542643.1
13
2979
XM_011544342.2
XP_011542644.1
12
3165
XM_011544343.2
XP_011542645.1
11
2688
XM_011544344.2
XP_011542646.1
12
2735
XM_017002932.1
XP_016858421.1
12
3128
XM_017002933.1
XP_016858422.1
13
2799
XM_017002934.1
XP_016858423.1
13
2442
XM_017002935.1
XP_016858424.1
13
2979
XM_017002936.1
XP_016858425.1
13
2442
XM_017002937.1
XP_016858426.1
13
2979
XM_017002938.1
XP_016858427.1
13
2979
XM_017002939.1
XP_016858428.1
13
2799
XM_017002940.1
XP_016858429.1
13
2442
XM_017002941.1
XP_016858430.1
13
2799
XM_017002942.1
XP_016858431.1
13
2979
XM_017002943.1
XP_016858432.1
13
2799
XM_017002944.1
XP_016858433.1
12
2728
XM_017002945.1
XP_016858434.1
12
2735
XM_017002946.1
XP_016858435.1
12
2502
XM_017002947.1
XP_016858436.1
12
2735
XM_017002948.1
XP_016858437.1
12
2728
XM_017002949.1
XP_016858438.1
12
2735
XM_017002950.1
XP_016858439.1
12
2502
XM_017002951.1
XP_016858440.1
11
2208
GenBank mRNA
AB007939.1
-
11
1850
AB022657.1
-
12
2144
AB022658.1
-
11
1850
AB022659.1
-
11
1850
AK296812.1
-
12
1923
AK308309.1
-
2
296
BC140794.1
-
11
1841
9021662
BC146790.1
-
11
1850
9007254
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-pfczg:80/100.66.78.229:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline