Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ROBO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145845.1 | NP_001139317.1 | 6, 8 | 1927 | ||
NM_133631.3 | NP_598334.2 | 6, 8 | 1927 | |||
XM_006713277.2 | XP_006713340.1 | 6, 8 | 1896 | |||
XM_011533976.1 | XP_011532278.1 | 8, 10 | 1343 | |||
XM_011533977.2 | XP_011532279.1 | 8, 10 | 1196 | |||
XM_011533978.1 | XP_011532280.1 | 8, 10 | 1343 | |||
XM_011533979.1 | XP_011532281.1 | 8, 10 | 1343 | |||
XM_011533980.1 | XP_011532282.1 | 8, 10 | 1343 | |||
XM_017006982.1 | XP_016862471.1 | 7, 9 | 1255 | |||
XM_017006985.1 | XP_016862474.1 | 6, 8 | 996 | |||
GenBank mRNA | AK095256.1 | - | 1, 3 | 862 | ||
BC112336.1 | - | 6, 8 | 980 | 40037251 | ||
BC115020.1 | - | 6, 8 | 1028 | 7961997 | ||
BC115022.1 | - | 6, 8 | 1028 | 7962000 | ||
BC157861.1 | - | 6, 8 | 1224 | 9057080 | ||
BC171855.1 | - | 6, 8 | 1224 | 9054509 |