Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA31C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145124.1 | NP_001138596.1 | 2 | 297 | ||
XM_011518701.2 | XP_011517003.1 | 3 | 935 | |||
XM_011518702.1 | XP_011517004.1 | 2 | 854 | |||
XM_011518703.1 | XP_011517005.1 | 2 | 734 | |||
GenBank mRNA | AK093374.1 | - | 4 | 994 | ||
DB065037.1 | - | 2 | 315 |
Gene Symbol: ANKRD19P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC028712.1 | - | 10 | 2149 |
Gene Symbol: LOC642943 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_111003.1 | 2 | 302 | |||
GenBank mRNA | BX097312.1 | - | 2 | 286 | ||
DB062721.1 | - | 2 | 302 | |||
HY033202.1 | - | 2 | 298 |
Gene Symbol: SPATA31A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085452.2 | NP_001078921.2 | 2 | 294 | ||
XM_011518006.1 | XP_011516308.1 | 2 | 332 | |||
GenBank mRNA | AK302569.1 | - | 2 | 294 | ||
BC146855.1 | - | 2 | 253 | 9056783 |
Gene Symbol: SPATA31A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083124.1 | NP_001076593.1 | 2 | 261 |
Gene Symbol: SPATA31A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113541.1 | NP_001107013.1 | 2 | 261 | ||
XM_011545671.2 | XP_011543973.1 | 2 | 332 |
Gene Symbol: SPATA31A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145196.1 | NP_001138668.1 | 2 | 261 | ||
XM_011517871.2 | XP_011516173.1 | 2 | 268 |
Gene Symbol: SPATA31A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015667.2 | NP_056482.2 | 2 | 261 | ||
GenBank mRNA | BC040528.1 | - | 2 | 261 | 4838470 | |
DB085566.1 | - | 2 | 83 |
Gene Symbol: SPATA31C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166137.1 | NP_001159609.1 | 2 | 279 | ||
GenBank mRNA | AK302448.1 | - | 2 | 279 |