Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00863 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029408.1 | 1 | 1031 |
Gene Symbol: LOC107985803 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017005446.1 | XP_016860935.1 | 5 | 2132 |
Gene Symbol: LOC642361 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029407.1 | 1 | 1492 | |||
GenBank mRNA | AF064843.1 | - | 1 | 1509 |