Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA31A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145196.1 | NP_001138668.1 | 4 | 3665 | ||
XM_011517871.2 | XP_011516173.1 | 4 | 3672 |