Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D22A-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_122047.1 | 1 | 569 | |||
GenBank mRNA | AK057318.1 | - | 1 | 547 | ||
BC041833.1 | - | 1 | 569 | |||
CR456602.1 | - | 1 | 435 |