Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABRR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256703.1 | NP_001243632.1 | 9 | 1752 | ||
NM_001256704.1 | NP_001243633.1 | 11 | 1905 | |||
NM_001267582.1 | NP_001254511.1 | 12 | 1573 | |||
NM_002042.4 | NP_002033.2 | 10 | 1803 | |||
XM_017010689.1 | XP_016866178.1 | 11 | 1393 | |||
GenBank mRNA | AK296124.1 | - | 9 | 1344 | ||
AK298948.1 | - | 12 | 1563 | |||
AK313609.1 | - | 11 | 1905 | |||
BC130344.1 | - | 10 | 1357 | 40146375 | ||
M62400.1 | - | 10 | 1376 |