Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMIM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162995.2 | NP_001156467.1 | 3 | 1188 | ||
NR_028439.1 | 2 | 3990 | ||||
XM_011525118.2 | XP_011523420.1 | 3 | 530 | |||
XM_011525119.2 | XP_011523421.1 | 3 | 438 | |||
XM_017024945.1 | XP_016880434.1 | 4 | 737 | |||
XM_017024946.1 | XP_016880435.1 | 3 | 1569 | |||
XM_017024947.1 | XP_016880436.1 | 3 | 687 | |||
XM_017024948.1 | XP_016880437.1 | 4 | 635 | |||
GenBank mRNA | AF370408.1 | - | 2 | 457 | ||
AK055768.1 | - | 2 | 2151 | |||
AL832643.2 | - | 2 | 3990 | |||
BC146918.1 | - | 2 | 1213 | |||
BC146923.1 | - | 2 | 1213 | |||
BI712658.1 | - | 3 | 551 |