Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIPK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080518.1 | NP_001073987.1 | 6 | 794 | ||
XM_011540073.2 | XP_011538375.1 | 7 | 895 | |||
XM_011540075.1 | XP_011538377.1 | 8 | 949 | |||
XM_011540077.1 | XP_011538379.1 | 7 | 886 | |||
XM_011540082.1 | XP_011538384.1 | 4 | 894 | |||
XM_017016545.1 | XP_016872034.1 | 7 | 895 | |||
XM_017016546.1 | XP_016872035.1 | 7 | 797 | |||
XM_017016548.1 | XP_016872037.1 | 7 | 898 | |||
GenBank mRNA | EF426482.1 | - | 6 | 794 |