Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000153.3 | NP_000144.2 | 4 | 460 | ||
NM_001201401.1 | NP_001188330.1 | 3 | 391 | |||
NM_001201402.1 | NP_001188331.1 | 4 | 441 | |||
XM_011536618.2 | XP_011534920.1 | 4 | 742 | |||
GenBank mRNA | AK294040.1 | - | 4 | 367 | ||
AK296530.1 | - | 3 | 282 | |||
AK302683.1 | - | 4 | 614 | |||
AK302956.1 | - | 4 | 280 | |||
BC036518.2 | - | 4 | 312 | 5267705 | ||
D25283.1 | - | 4 | 343 | |||
D25284.1 | - | 4 | 343 | |||
DA247447.1 | - | 4 | 384 | |||
DA332567.1 | - | 4 | 424 | |||
DC334180.1 | - | 4 | 441 | |||
L23116.1 | - | 4 | 352 |