Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNPS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286625.1 | NP_001273554.1 | 6, 7 | 957 | ||
NM_001286626.1 | NP_001273555.1 | 5, 6 | 964 | |||
NM_006711.4 | NP_006702.1 | 6, 7 | 1018 | |||
NM_080594.3 | NP_542161.1 | 6, 7 | 1152 | |||
NR_104485.1 | 5, 6 | 1086 | ||||
XM_005255048.1 | XP_005255105.1 | 6, 7 | 1245 | |||
XM_005255049.3 | XP_005255106.1 | 6, 7 | 2710 | |||
XM_017022874.1 | XP_016878363.1 | 6, 7 | 1611 | |||
GenBank mRNA | AB445101.1 | - | 5, 6 | 677 | ||
AF015608.1 | - | 5, 6 | 691 | |||
AF247662.1 | - | 3, 4 | 446 | |||
AF274003.1 | - | 5, 6 | 653 | |||
AK289955.1 | - | 7, 8 | 912 | |||
AK293343.1 | - | 5, 6 | 669 | |||
AK303100.1 | - | 5, 6 | 625 | |||
AK316132.1 | - | 5, 6 | 954 | |||
AL559988.3 | - | 5, 6 | 738 | |||
BC001659.2 | - | 6, 7 | 842 | 2959954 | ||
BC001838.2 | - | 6, 7 | 842 | 2959954 | ||
BC108316.1 | - | 6, 7 | 963 | 6151864 | ||
BQ059026.1 | - | 2 | 820 | |||
L37368.1 | - | 6, 7 | 1226 |