Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHFR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000791.3 | NP_000782.1 | 4 | 804 | ||
NM_001290354.1 | NP_001277283.1 | 3 | 754 | |||
NM_001290357.1 | NP_001277286.1 | 4 | 804 | |||
GenBank mRNA | AK293146.1 | - | 3 | 332 | ||
AK297302.1 | - | 4 | 375 | |||
AK312642.1 | - | 1 | 348 | |||
BC000192.1 | - | 4 | 367 | 3352539 | ||
BC003584.1 | - | 5 | 766 | 3354190 | ||
BC070280.1 | - | 4 | 349 | 6714220 | ||
BC071996.1 | - | 5 | 766 | 4645596 | ||
EU145592.1 | - | 1 | 312 | |||
J00140.1 | - | 4 | 354 | |||
V00507.1 | - | 1 | 312 |