Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000403.3 | NP_000394.2 | 7 | 849 | ||
NM_001008216.1 | NP_001008217.1 | 7 | 784 | |||
NM_001127621.1 | NP_001121093.1 | 6 | 825 | |||
GenBank mRNA | AK057302.1 | - | 5 | 1020 | ||
AK314397.1 | - | 6 | 797 | |||
AL530475.3 | - | 7 | 783 | |||
BC001273.1 | - | 6 | 705 | 3459004 | ||
BC050685.2 | - | 7 | 708 | 6161561 | ||
DQ233668.1 | - | 5 | 615 | |||
L41668.1 | - | 7 | 708 |