Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERICH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290030.1 | NP_001276959.1 | 3 | 574 | ||
NM_001290031.1 | NP_001276960.1 | 3 | 574 | |||
NR_110644.1 | 4 | 881 | ||||
XM_011510998.2 | XP_011509300.1 | 5 | 706 | |||
XM_011510999.2 | XP_011509301.1 | 4 | 373 | |||
XM_011511001.2 | XP_011509303.1 | 3 | 1052 | |||
XM_017003875.1 | XP_016859364.1 | 4 | 1075 | |||
XM_017003876.1 | XP_016859365.1 | 3 | 90 | |||
XM_017003877.1 | XP_016859366.1 | 4 | 373 | |||
GenBank mRNA | AA401652.1 | - | 3 | 125 | ||
BG215457.1 | - | 1 | 75 | |||
BQ269226.1 | - | 4 | 450 |