Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001556.2 | NP_001001556.1 | 9 | 1234 | ||
NM_001289030.1 | NP_001275959.1 | 10 | 1466 | |||
NM_001289031.1 | NP_001275960.1 | 9 | 1319 | |||
NM_002044.3 | NP_002035.1 | 9 | 1336 | |||
XM_005254279.3 | XP_005254336.1 | 9 | 1125 | |||
XM_005254280.3 | XP_005254337.1 | 9 | 1216 | |||
XM_005254284.3 | XP_005254341.1 | 8 | 1117 | |||
XM_006720461.3 | XP_006720524.1 | 9 | 1125 | |||
XM_006720462.3 | XP_006720525.1 | 9 | 1106 | |||
XM_006720463.3 | XP_006720526.1 | 8 | 1006 | |||
XM_006720464.3 | XP_006720527.1 | 7 | 853 | |||
XM_011521441.2 | XP_011519743.1 | 10 | 1329 | |||
XM_017022062.1 | XP_016877551.1 | 8 | 1007 | |||
XM_017022063.1 | XP_016877552.1 | 7 | 964 | |||
XM_017022064.1 | XP_016877553.1 | 7 | 1182 | |||
XM_017022065.1 | XP_016877554.1 | 10 | 1381 | |||
XM_017022066.1 | XP_016877555.1 | 10 | 1402 | |||
GenBank mRNA | AF461816.1 | - | 9 | 1214 | ||
AK098049.1 | - | 11 | 1335 | |||
AK301361.1 | - | 9 | 1089 | |||
AK316440.1 | - | 10 | 1200 | |||
BC005141.2 | - | 9 | 1104 | 3504930 | ||
BT006901.1 | - | 9 | 1029 | |||
M84443.1 | - | 9 | 1049 |