Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRNDE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308963.1 | NP_001295892.1 | 5 | 509 | ||
NR_034105.3 | 5 | 727 | ||||
NR_034106.2 | 5 | 592 | ||||
NR_110453.1 | 6 | 814 | ||||
NR_110454.1 | 5 | 520 | ||||
XM_017023562.1 | XP_016879051.1 | 5 | 341 | |||
XM_017023563.1 | XP_016879052.1 | 5 | 1642 | |||
GenBank mRNA | BU180741.1 | - | 5 | 359 | ||
CV575277.1 | - | 5 | 458 | |||
DB446128.1 | - | 6 | 435 | |||
FJ466685.1 | - | 5 | 365 | |||
FJ466686.1 | - | 5 | 282 |