Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRNDE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308963.1 | NP_001295892.1 | 5 | 538 | ||
NR_034105.3 | 5 | 756 | ||||
NR_034106.2 | 5 | 621 | ||||
NR_110453.1 | 6 | 843 | ||||
NR_110454.1 | 5 | 549 | ||||
XM_017023562.1 | XP_016879051.1 | 5 | 370 | |||
XM_017023563.1 | XP_016879052.1 | 5 | 1671 | |||
GenBank mRNA | AF275804.1 | - | 5 | 364 | ||
BF692451.1 | - | 5 | 627 | |||
BU180741.1 | - | 5 | 388 | |||
CV575277.1 | - | 5 | 487 | |||
FJ466685.1 | - | 5 | 394 | |||
FJ466686.1 | - | 5 | 311 |