Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291866.1 | NP_001278795.1 | 10 | 1098 | ||
NM_004481.4 | NP_004472.1 | 10 | 1020 | |||
XM_017000963.1 | XP_016856452.1 | 10 | 980 | |||
XM_017000964.1 | XP_016856453.1 | 10 | 1605 | |||
XM_017000965.1 | XP_016856454.1 | 10 | 1098 | |||
XM_017000966.1 | XP_016856455.1 | 10 | 923 | |||
GenBank mRNA | AK290048.1 | - | 10 | 946 | ||
AK296886.1 | - | 7 | 617 | |||
AK300453.1 | - | 10 | 1098 | |||
BC041120.1 | - | 10 | 1006 | 5553465 | ||
X85019.1 | - | 10 | 934 |