Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291866.1 | NP_001278795.1 | 13 | 1440 | ||
NM_004481.4 | NP_004472.1 | 13 | 1362 | |||
XM_017000963.1 | XP_016856452.1 | 13 | 1322 | |||
XM_017000964.1 | XP_016856453.1 | 13 | 1947 | |||
XM_017000965.1 | XP_016856454.1 | 13 | 1440 | |||
XM_017000966.1 | XP_016856455.1 | 13 | 1265 | |||
GenBank mRNA | AK097996.1 | - | 2 | 761 | ||
AK290048.1 | - | 13 | 1288 | |||
AK296886.1 | - | 10 | 959 | |||
AK300453.1 | - | 13 | 1440 | |||
AK304029.1 | - | 11 | 1088 | |||
BC041120.1 | - | 13 | 1348 | 5553465 | ||
BC050583.1 | - | 3 | 928 | |||
X85019.1 | - | 13 | 1276 |