Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPN8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143962.1 | NP_001137434.1 | 17, 18 | 1931 | ||
XM_017001265.1 | XP_016856754.1 | 14, 15 | 1372 | |||
XM_017001266.1 | XP_016856755.1 | 12, 13 | 1289 | |||
GenBank mRNA | BC157893.1 | - | 17, 18 | 1868 | 9057098 | |
BM843240.1 | - | 1, 2 | 76 |