Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000155.3 | NP_000146.2 | 3 | 391 | ||
GenBank mRNA | AK096137.1 | - | 3 | 305 | ||
AK293212.1 | - | 3 | 334 | |||
AK300070.1 | - | 3 | 280 | |||
AK314340.1 | - | 3 | 373 | |||
BC015045.2 | - | 3 | 316 | 3922902 | ||
BT009852.1 | - | 3 | 275 | |||
KU178089.1 | - | 3 | 275 | |||
KU178090.1 | - | 3 | 275 | |||
M60091.1 | - | 3 | 303 |