Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC107987253 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017025512.1 | XP_016881001.1 | 1 | 471 |
Gene Symbol: FAM215A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026770.1 | 1 | 506 | |||
GenBank mRNA | AF143236.1 | - | 1 | 518 | ||
BC069097.1 | - | 1 | 506 |
Gene Symbol: LOC107987254 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017025513.1 | XP_016881002.1 | 1 | 471 |