Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MT1IP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003669.2 | 3 | 322 | |||
NR_104045.1 | 3 | 307 | ||||
NR_104046.1 | 2 | 278 | ||||
GenBank mRNA | AF348997.1 | - | 3 | 345 |