Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRTAP2-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033184.3 | NP_149440.1 | 1 | 620 | ||
GenBank mRNA | AJ406930.1 | - | 1 | 620 | ||
BC101147.2 | - | 1 | 616 | 40015180 | ||
BC101148.2 | - | 1 | 616 | 40015187 | ||
BC101149.2 | - | 1 | 616 | 40015188 |