Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAPDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256799.2 | NP_001243728.1 | 5 | 639 | ||
NM_001289745.1 | NP_001276674.1 | 6 | 697 | |||
NM_001289746.1 | NP_001276675.1 | 5 | 591 | |||
NM_002046.5 | NP_002037.2 | 6 | 605 | |||
GenBank mRNA | AB062273.1 | - | 6 | 469 | ||
AF261085.1 | - | 6 | 520 | |||
AK026525.1 | - | 6 | 542 | |||
AK299448.1 | - | 6 | 490 | |||
AK308198.1 | - | 6 | 493 | |||
AY007133.1 | - | 6 | 469 | |||
AY633612.1 | - | 5 | 441 | |||
BC001601.1 | - | 6 | 471 | 3543589 | ||
BC004109.2 | - | 6 | 469 | 3678259 | ||
BC009081.1 | - | 6 | 478 | 3869809 | ||
BC013310.2 | - | 6 | 477 | 3538383 | ||
BC020308.1 | - | 5 | 455 | |||
BC023632.2 | - | 5 | 441 | 4842418 | ||
BC025925.1 | - | 6 | 474 | 2989348 | ||
BC026907.1 | - | 5 | 455 | 4454083 | ||
BC029340.1 | - | 6 | 463 | |||
BC029618.1 | - | 6 | 471 | 3876535 | ||
BC083511.1 | - | 6 | 469 | 3536969 | ||
BG724119.1 | - | 5 | 623 | |||
BM763361.1 | - | 6 | 582 | |||
BT006893.1 | - | 5 | 417 | |||
BU155402.1 | - | 6 | 642 | |||
CR407671.1 | - | 5 | 417 | |||
DQ403057.1 | - | 4 | 342 | |||
EF036498.1 | - | 2 | 155 | |||
EU668321.1 | - | 6 | 519 | |||
J02642.1 | - | 5 | 418 | |||
M17851.1 | - | 5 | 450 | |||
M33197.1 | - | 6 | 477 | |||
X01677.1 | - | 5 | 477 | |||
X53778.1 | - | 6 | 473 |