Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAPK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001348.2 | NP_001339.1 | 7 | 985 | ||
XM_005259508.1 | XP_005259565.1 | 7 | 860 | |||
GenBank mRNA | AB007144.1 | - | 6 | 781 | ||
AB022341.1 | - | 6 | 733 | |||
AB265224.1 | - | 4 | 198 | |||
AK027590.1 | - | 7 | 902 | |||
AK074799.1 | - | 6 | 757 | |||
AK093792.1 | - | 1 | 738 | |||
AK097643.1 | - | 7 | 796 | |||
BC126430.1 | - | 6 | 724 | 8992146 | ||
BC126432.1 | - | 6 | 722 | 8992148 |