Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAS8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286205.1 | NP_001273134.1 | 9 | 1344 | ||
NM_001286208.1 | NP_001273137.1 | 8 | 1289 | |||
NM_001286209.1 | NP_001273138.1 | 9 | 1236 | |||
NM_001481.2 | NP_001472.1 | 9 | 1221 | |||
XM_005256304.4 | XP_005256361.1 | 10 | 1876 | |||
XM_005256309.4 | XP_005256366.1 | 11 | 1793 | |||
XM_006721175.3 | XP_006721238.1 | 9 | 1274 | |||
XM_011522990.2 | XP_011521292.1 | 9 | 1295 | |||
XM_011522991.1 | XP_011521293.1 | 10 | 1450 | |||
XM_011522992.2 | XP_011521294.1 | 8 | 1243 | |||
XM_017023122.1 | XP_016878611.1 | 11 | 1601 | |||
XM_017023123.1 | XP_016878612.1 | 9 | 1300 | |||
XM_017023124.1 | XP_016878613.1 | 8 | 1087 | |||
XM_017023125.1 | XP_016878614.1 | 8 | 1219 | |||
GenBank mRNA | AF050079.1 | - | 9 | 1220 | ||
AK293958.1 | - | 9 | 1251 | |||
AK294050.1 | - | 8 | 1070 | |||
AK297857.1 | - | 9 | 1204 | |||
AK315261.1 | - | 9 | 1169 | |||
BC104785.1 | - | 9 | 1206 | 8143788 | ||
BC112121.1 | - | 9 | 1206 | 8327589 | ||
BX647947.1 | - | 8 | 1207 | |||
CR749499.1 | - | 7 | 1899 |