Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAS8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286205.1 | NP_001273134.1 | 5 | 768 | ||
NM_001286209.1 | NP_001273138.1 | 5 | 660 | |||
NM_001481.2 | NP_001472.1 | 5 | 645 | |||
XM_005256304.4 | XP_005256361.1 | 6 | 1300 | |||
XM_005256309.4 | XP_005256366.1 | 7 | 1025 | |||
XM_006721175.3 | XP_006721238.1 | 5 | 698 | |||
XM_011522990.2 | XP_011521292.1 | 5 | 719 | |||
XM_011522991.1 | XP_011521293.1 | 6 | 874 | |||
XM_011522992.2 | XP_011521294.1 | 4 | 667 | |||
XM_017023122.1 | XP_016878611.1 | 7 | 1025 | |||
XM_017023123.1 | XP_016878612.1 | 5 | 724 | |||
GenBank mRNA | AF050079.1 | - | 5 | 644 | ||
AK293958.1 | - | 5 | 675 | |||
AK294050.1 | - | 4 | 494 | |||
AK297857.1 | - | 5 | 628 | |||
AK303505.1 | - | 5 | 701 | |||
AK315261.1 | - | 5 | 593 | |||
BC104785.1 | - | 5 | 630 | 8143788 | ||
BC112121.1 | - | 5 | 630 | 8327589 | ||
CR749499.1 | - | 4 | 678 |