Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLTP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016433.3 | NP_057517.1 | 1, 2 | 215 | ||
XM_011538434.2 | XP_011536736.1 | 1, 2 | 168 | |||
GenBank mRNA | AF209704.1 | - | 1, 2 | 111 | ||
AK313457.1 | - | 1, 2 | 104 | |||
AY372530.1 | - | 1, 2 | 101 | |||
AY372531.1 | - | 1, 2 | 101 | |||
AY372532.1 | - | 1, 2 | 130 | |||
BC009932.2 | - | 1, 2 | 215 | 3959556 | ||
JN039379.1 | - | 1, 2 | 101 |