Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GATA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308093.1 | NP_001295022.1 | 2 | 1158 | ||
NM_001308094.1 | NP_001295023.1 | 3 | 498 | |||
NM_002052.4 | NP_002043.2 | 3 | 1309 | |||
XM_005272385.4 | XP_005272442.1 | 4 | 1381 | |||
XM_005272386.1 | XP_005272443.1 | 4 | 1569 | |||
XM_006716248.1 | XP_006716311.1 | 4 | 1490 | |||
XM_011543817.2 | XP_011542119.1 | 4 | 1560 | |||
XM_011543818.2 | XP_011542120.1 | 3 | 1698 | |||
XM_017013312.1 | XP_016868801.1 | 3 | 2014 | |||
GenBank mRNA | AK097060.1 | - | 3 | 458 | ||
AY740706.1 | - | 2 | 701 | |||
BC101580.1 | - | 2 | 778 | 8069086 | ||
BC105108.1 | - | 2 | 776 | 8144111 | ||
BC143434.1 | - | 2 | 778 | 9051942 | ||
BC143479.1 | - | 2 | 781 | 9051987 | ||
D78260.1 | - | 2 | 990 | |||
DA900813.1 | - | 2 | 239 | |||
L34357.1 | - | 2 | 938 |