Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GATA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308093.1 | NP_001295022.1 | 4 | 1386 | ||
NM_001308094.1 | NP_001295023.1 | 5 | 726 | |||
NM_002052.4 | NP_002043.2 | 5 | 1537 | |||
XM_005272385.4 | XP_005272442.1 | 6 | 1609 | |||
XM_005272386.1 | XP_005272443.1 | 6 | 1797 | |||
XM_006716248.1 | XP_006716311.1 | 6 | 1718 | |||
XM_011543817.2 | XP_011542119.1 | 6 | 1788 | |||
XM_011543818.2 | XP_011542120.1 | 5 | 1926 | |||
XM_017013312.1 | XP_016868801.1 | 5 | 2242 | |||
GenBank mRNA | AK097060.1 | - | 5 | 686 | ||
AY740706.1 | - | 4 | 929 | |||
BC101580.1 | - | 4 | 1006 | 8069086 | ||
BC105108.1 | - | 4 | 1004 | 8144111 | ||
BC143434.1 | - | 4 | 1006 | 9051942 | ||
BC143479.1 | - | 4 | 1009 | 9051987 | ||
D78260.1 | - | 4 | 1218 | |||
DA900813.1 | - | 4 | 467 | |||
L34357.1 | - | 4 | 1166 |