Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC107133515 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015341.2 | 1, 2 | 204 | |||
GenBank mRNA | BG721507.1 | - | 1, 2 | 165 | ||
DB024747.1 | - | 1, 2 | 204 |