Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2E1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202476.1 | NP_001189405.1 | 3 | 222 | ||
NM_003341.4 | NP_003332.1 | 4 | 517 | |||
NM_182666.2 | NP_872607.1 | 3 | 466 | |||
XM_005265431.4 | XP_005265488.1 | 3 | 274 | |||
GenBank mRNA | AK295354.1 | - | 3 | 222 | ||
AK314854.1 | - | 4 | 426 | |||
BC009139.1 | - | 4 | 441 | 3853408 | ||
BI223271.1 | - | 3 | 361 | |||
DB499676.1 | - | 3 | 222 | |||
X92963.1 | - | 3 | 288 |