Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GBAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001483.2 | NP_001474.1 | 3 | 296 | ||
GenBank mRNA | AF029786.1 | - | 3 | 271 | ||
AJ001259.1 | - | 3 | 263 | |||
AK315648.1 | - | 3 | 281 | |||
BC000732.2 | - | 3 | 269 | 2959662 | ||
BC001837.2 | - | 3 | 269 | 2959662 | ||
BC030821.1 | - | 3 | 278 | 4808962 | ||
BT007112.1 | - | 3 | 263 | |||
CR407612.1 | - | 3 | 263 | |||
CX166452.1 | - | 3 | 296 |