Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP17L7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256869.1 | NP_001243798.1 | 1 | 129 |
Gene Symbol: USP17L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256873.1 | NP_001243802.1 | 1 | 129 |
Gene Symbol: USP17L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256871.1 | NP_001243800.1 | 1 | 129 |
Gene Symbol: USP17L4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256874.1 | NP_001243803.1 | 1 | 129 |
Gene Symbol: USP17L8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256872.1 | NP_001243801.1 | 1 | 129 |