Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004120.4 | NP_004111.2 | 2 | 358 | ||
GenBank mRNA | AK057968.1 | - | 2 | 271 | ||
AK226018.1 | - | 2 | 284 | |||
AK314325.1 | - | 2 | 313 | |||
BC022272.1 | - | 2 | 313 | 4688776 | ||
BC073163.1 | - | 2 | 295 | 6151056 | ||
CA390345.1 | - | 2 | 341 | |||
CR457062.1 | - | 1 | 44 | |||
DB274428.1 | - | 2 | 358 | |||
M55543.1 | - | 2 | 200 |