Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319179.1 | NP_001306108.1 | 8 | 1177 | ||
NM_001319180.1 | NP_001306109.1 | 7 | 1233 | |||
NM_001319181.1 | NP_001306110.1 | 7 | 1091 | |||
NM_018284.2 | NP_060754.2 | 7 | 1091 | |||
XM_005270743.2 | XP_005270800.1 | 8 | 1167 | |||
XM_006710574.2 | XP_006710637.1 | 7 | 1105 | |||
XM_011541233.1 | XP_011539535.1 | 6 | 879 | |||
XM_011541234.1 | XP_011539536.1 | 6 | 862 | |||
XM_017001013.1 | XP_016856502.1 | 6 | 1011 | |||
XM_017001014.1 | XP_016856503.1 | 5 | 759 | |||
GenBank mRNA | AK001823.1 | - | 7 | 1205 | ||
AK309096.1 | - | 6 | 1731 | |||
AL136680.1 | - | 7 | 990 | |||
BC017992.1 | - | 7 | 1063 | |||
BC140837.1 | - | 7 | 976 | 9021705 | ||
BC144650.1 | - | 8 | 1062 | |||
CR936755.1 | - | 7 | 1092 | |||
CR936758.1 | - | 7 | 1032 | |||
JF927951.1 | - | 7 | 955 |