Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319179.1 | NP_001306108.1 | 11 | 1937 | ||
NM_001319180.1 | NP_001306109.1 | 10 | 1993 | |||
NM_001319181.1 | NP_001306110.1 | 10 | 1770 | |||
NM_018284.2 | NP_060754.2 | 10 | 1851 | |||
NR_135007.1 | 8 | 1357 | ||||
XM_005270743.2 | XP_005270800.1 | 11 | 1927 | |||
XM_006710574.2 | XP_006710637.1 | 10 | 1784 | |||
XM_011541233.1 | XP_011539535.1 | 9 | 1639 | |||
XM_011541234.1 | XP_011539536.1 | 9 | 1622 | |||
XM_017001013.1 | XP_016856502.1 | 9 | 1771 | |||
XM_017001014.1 | XP_016856503.1 | 8 | 1519 | |||
GenBank mRNA | AK001823.1 | - | 10 | 1965 | ||
AK309096.1 | - | 9 | 2491 | |||
AL136680.1 | - | 10 | 1750 | |||
BC063819.1 | - | 8 | 1269 | |||
BC140837.1 | - | 10 | 1736 | 9021705 | ||
BC144650.1 | - | 11 | 1822 | |||
CR936755.1 | - | 10 | 1852 | |||
CR936758.1 | - | 10 | 1792 | |||
JF927951.1 | - | 10 | 1634 |