Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319179.1 | NP_001306108.1 | 11, 12 | 1959 | ||
NM_001319180.1 | NP_001306109.1 | 10, 11 | 2015 | |||
NM_018284.2 | NP_060754.2 | 10, 11 | 1873 | |||
NR_135007.1 | 8, 9 | 1379 | ||||
XM_005270743.2 | XP_005270800.1 | 11, 12 | 1949 | |||
XM_006710574.2 | XP_006710637.1 | 10, 11 | 1806 | |||
XM_011541233.1 | XP_011539535.1 | 9, 10 | 1661 | |||
XM_011541234.1 | XP_011539536.1 | 9, 10 | 1644 | |||
XM_017001013.1 | XP_016856502.1 | 9, 10 | 1793 | |||
XM_017001014.1 | XP_016856503.1 | 8, 9 | 1541 | |||
GenBank mRNA | AK001823.1 | - | 10, 11 | 1987 | ||
AK309096.1 | - | 9, 10 | 2513 | |||
AL136680.1 | - | 10, 11 | 1772 | |||
BC063819.1 | - | 8, 9 | 1291 | |||
BC140837.1 | - | 10, 11 | 1758 | 9021705 | ||
BC144650.1 | - | 11, 12 | 1844 | |||
CR936755.1 | - | 10, 11 | 1874 | |||
CR936758.1 | - | 10, 11 | 1814 |