Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100132062 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028325.1 | 3 | 1790 | |||
GenBank mRNA | AK303380.1 | - | 1 | 517 | ||
AK316554.1 | - | 1 | 679 | |||
AK316556.1 | - | 1 | 677 | |||
BC112924.1 | - | 1 | 1178 |
Gene Symbol: LINC01001 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028326.1 | 3 | 1790 |
Gene Symbol: LOC100132287 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028322.1 | 3 | 1790 | |||
GenBank mRNA | AK311126.1 | - | 1 | 964 |
Gene Symbol: LOC100133331 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028327.1 | 3 | 1790 |
Gene Symbol: LOC729737 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_039983.2 | 3 | 2113 | |||
GenBank mRNA | AK294489.1 | - | 1 | 1446 | ||
AK302573.1 | - | 1 | 1076 |