Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GCDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000159.3 | NP_000150.1 | 7 | 702 | ||
NM_013976.3 | NP_039663.1 | 7 | 702 | |||
NR_102316.1 | 7 | 757 | ||||
NR_102317.1 | 6 | 1010 | ||||
XM_006722721.2 | XP_006722784.1 | 7 | 671 | |||
XM_011527899.2 | XP_011526201.1 | 7 | 671 | |||
XM_011527900.2 | XP_011526202.1 | 7 | 671 | |||
XM_017026580.1 | XP_016882069.1 | 7 | 671 | |||
GenBank mRNA | AK098370.1 | - | 7 | 671 | ||
AK290407.1 | - | 7 | 671 | |||
AK296446.1 | - | 7 | 720 | |||
AK298772.1 | - | 5 | 572 | |||
AK300841.1 | - | 6 | 959 | |||
AU135567.1 | - | 7 | 671 | |||
BC002579.2 | - | 7 | 655 | 3142978 | ||
BT006706.1 | - | 6 | 594 | |||
U69141.1 | - | 6 | 630 |