Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202498.1 | NP_001189427.1 | 5, 6 | 303 | ||
NM_003344.3 | NP_003335.1 | 5, 6 | 699 | |||
GenBank mRNA | AB451490.1 | - | 5, 6 | 294 | ||
AK316590.1 | - | 5, 6 | 282 | |||
BC006277.2 | - | 5, 6 | 611 | 3957107 | ||
BI088146.1 | - | 5, 6 | 303 | |||
BT006756.1 | - | 5, 6 | 294 | |||
KJ534959.1 | - | 5, 6 | 294 | |||
Z29328.1 | - | 5, 6 | 347 | |||
Z29330.1 | - | 5, 6 | 347 |