Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYPD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085474.1 | NP_001078943.1 | 5 | 466 | ||
NM_001291283.1 | NP_001278212.1 | 4 | 318 | |||
GenBank mRNA | AY358469.1 | - | 5 | 418 | ||
DA425843.1 | - | 4 | 283 | |||
DA922659.1 | - | 4 | 319 |