Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYPD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085474.1 | NP_001078943.1 | 6, 7 | 758 | ||
NM_001291283.1 | NP_001278212.1 | 5, 6 | 610 | |||
GenBank mRNA | AY358469.1 | - | 6, 7 | 710 | ||
DA425843.1 | - | 575 |