Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GCK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000162.3 | NP_000153.1 | 4 | 942 | ||
NM_033507.1 | NP_277042.1 | 4 | 643 | |||
NM_033508.1 | NP_277043.1 | 5 | 767 | |||
XM_017011966.1 | XP_016867455.1 | 4 | 958 | |||
GenBank mRNA | AK122876.1 | - | 4 | 637 | ||
BC001890.1 | - | 4 | 942 | 3536582 | ||
BT007062.1 | - | 4 | 472 | |||
EU008605.1 | - | 4 | 472 | |||
M69051.1 | - | 5 | 754 | |||
M88011.1 | - | 4 | 801 | |||
M90299.1 | - | 4 | 930 |